

1,8-Diaminooctane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:373-44-4
:
Formula:C₈H₂₀N₂
MDL Number:MFCD00008248
:144.26 g/mol
Boiling Pt:240°C
Melting Pt:51 °C
Density:0,83 g/cm³ (60 °C)
Storage Temperature:Ambient
EINECS:206-764-3
UN:3259
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together