
1,7-Diaminoheptane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:646-19-5
:
Formula:C₇H₁₈N₂
MDL Number:MFCD00008246
:130.23 g/mol
Boiling Pt:225°C
Melting Pt:28…29 °C
Density:1,012 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:211-468-2
UN:3259
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together