
1,6-Dihydroxynaphthalene ≥99.0% (by GC)
: TCI
:575-44-0
:
:C₁₀H₈O₂
:MFCD00003981
:160.17 g/mol
:375 °C (1013 hPa)
:135.5 °C
:Ambient
:209-386-7
:GHS07
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:135.0 to 139.0 deg-C
- Solubility in Methanol:very faint turbidity
Frequently Bought Together