1,4-Dimethoxy-2-nitrobenzene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:89-39-4
:
Formula:C₈H₉NO₄
MDL Number:MFCD00024211
:183.16 g/mol
Boiling Pt:169°C
EINECS:201-903-4
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:70.0 to 73.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together