
1,4-Bis(1,2-dibromoethyl)benzene ≥97.0% (by GC, titration analysis)
Catalog # TCIAB1504-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:157.0 to 162.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
About this item
:
:449.81 g/mol
EINECS:246-936-5
UN:1759
ADR:8,III
:GHS05
CAS Number:25393-98-0
Formula:C₁₀H₁₀Br₄
MDL Number:MFCD00017881