
Succinic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:110-15-6
:
Formula:HOOCCH₂CH₂COOH
MDL Number:MFCD00002789
:118.09 g/mol
Merck Index:14,08869
Boiling Pt:235 °C (1013 hPa)
Melting Pt:182 °C
Density:1,56 g/cm³ (20 °C)
Flash Pt:206 °C
Storage Temperature:Ambient
EINECS:203-740-4
ADR:8,III
:GHS07
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:185.0 to 189.0 deg-C
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together