Order Entry
Netherlands
ContactUsLinkComponent
5-Ethyl-m-xylene ≥98.0% (by GC)
Flammable
5-Ethyl-m-xylene ≥98.0% (by GC)
Catalog # TCIAE0282-0.1ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
5-Ethyl-m-xylene ≥98.0% (by GC)
Catalog # TCIAE0282-0.1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    0,1 mL
  • Seq:
    2010

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :134.22 g/mol
Boiling Pt:184°C
Melting Pt:-84°C
Melting Pt:–84°C
Flash Pt:52°C
EINECS:213-290-0
UN:3295
ADR:3,III
 :GHS02
CAS Number:934-74-7
Formula:C₁₀H₁₄
MDL Number:MFCD00059235