
2,6-Dimethylphenyl isocyanate ≥98.0% (by GC)
: TCI
:28556-81-2
:
:C₉H₉NO
:MFCD00002012
:147.18 g/mol
:113°C
:1,057 g/cm³ (20 °C)
:86°C
:Refrigerator
:2206
:6.1,III
:GHS06
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together