Order Entry
Netherlands
ContactUsLinkComponent
2,4-Difluorophenyl isothiocyanate ≥98.0% (by GC)
Giftig
2,4-Difluorophenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAD3454-1G
Leverancier:  TCI
CAS-nummer:  
undefined
Giftig
2,4-Difluorophenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAD3454-1G
Leverancier:  TCI
CAS-nummer:  

Specificaties

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specificaties

Specificatie testresultaten

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Refractive index n20/D:
    1.5940 to 1.5980

Specificatie testresultaten

Over dit artikel

 :
 :171.17 g/mol
Kokend Pt:86°C
Dichtheid:1,349 g/cm³ (20 °C)
UN:2810
ADR:6.1,III
 :GHS06
CAS-nummer:141106-52-7
Formule:C₇H₃F₂NS
MDL-nummer:MFCD00041044