Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
2,4-Difluorophenyl isocyanate ≥98.0% (by GC)
  TCIAD3086-5G
 :  TCI
 :  
undefined
 
 
 
2,4-Difluorophenyl isocyanate ≥98.0% (by GC)
  TCIAD3086-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :50°C
 :1,3 g/cm³ (20 °C)
 :55°C
 :3,III
 :155.1 g/mol
 :2478
 :GHS02
 :GHS06
 :GHS08
 :Refrigerator
 :59025-55-7
 :C₇H₃F₂NO
 :MFCD00001997