Order Entry
Netherlands
ContactUsLinkComponent
 
 
3,5-Dichlorophenyl isocyanate ≥97.0% (by GC, titration analysis)
  TCIAI0302-10G
 :  TCI
 :  
undefined
 
3,5-Dichlorophenyl isocyanate ≥97.0% (by GC, titration analysis)
  TCIAI0302-10G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 97.0 %
  • Purity (Butylamine Method):
    min. 97.0 %

 

 

 :
 :188.01 g/mol
 :264 °C (1013 hPa)
 :32…34 °C
 :1,38 g/cm³ (20 °C)
 :Refrigerator
 :252-276-9
 :2250
 :6.1,II
 :GHS06
 :34893-92-0
 :C₇H₃Cl₂NO
 :MFCD00013859