
α,α,α',α'-Tetrabromo-m-xylene ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:36323-28-1
:
Formula:C₈H₆Br₄
MDL Number:MFCD00000132
:421.75 g/mol
EINECS:252-975-9
UN:1759
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:105.0 to 109.0 deg-C
- Solubility in Toluene:almost transparency
Specification Test Results
Frequently Bought Together