

α,α'-Dichloro-o-xylene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:612-12-4
:
Formula:C₈H₈Cl₂
MDL Number:MFCD00000903
:175.06 g/mol
Boiling Pt:241°C
Melting Pt:54…55 °C
Density:1,393 g/cm³ (0 °C)
EINECS:210-291-8
UN:2928
ADR:6.1,II
:GHS05
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:55.0 to 57.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together