Order Entry
Netherlands
ContactUsLinkComponent
L(+)-Glutamic acid ≥99.0% (by titrimetric analysis)
L(+)-Glutamic acid ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
L(+)-Glutamic acid ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Specific rotation [a]20/D:
    +31.5 to +32.5 deg (C=5, HCl (1+5))
  • Solubility in dilute HCl:
    almost transparency

Specification Test Results

Frequently Bought Together