

(R)-(-)-2-Pentanol ≥98.0% (by GC)
: TCI
:31087-44-2
:
:C₅H₁₂O
:MFCD00065953
:88.15 g/mol
:119°C
:34°C
:Ambient
:1105
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:-12.0 to -15.0 deg (neat)
Frequently Bought Together