Order Entry
Netherlands
ContactUsLinkComponent
4-Chlorobutyl phenyl ether ≥95.0% (by GC)
4-Chlorobutyl phenyl ether ≥95.0% (by GC)
Catalog # TCIAP1386-25G
Supplier:  TCI
CAS Number:  
undefined
4-Chlorobutyl phenyl ether ≥95.0% (by GC)
Catalog # TCIAP1386-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :184.67 g/mol
Boiling Pt:144°C
Melting Pt:20 °C
EINECS:220-174-3
CAS Number:2651-46-9
Formula:(C₆H₅)O(CH₂)₄Cl
MDL Number:MFCD00001012