
(4-Bromophenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1878-68-8
:
Formula:(C₆H₄Br)CH₂COOH
MDL Number:MFCD00004342
:215.05 g/mol
Boiling Pt:326 °C (1013 hPa)
Melting Pt:94…98 °C
EINECS:217-523-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:115.0 to 121.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together