
(1-Hexyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4762-26-9
:
Formula:H₃C(CH₂)₅P(C₆H₅)₃Br
MDL Number:MFCD00011860
:427.36 g/mol
Melting Pt:198…200 °C
EINECS:225-301-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:199.0 to 204.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together