

1-Methylpiperidine ≥99.0% (by GC, titration analysis)
: TCI
:626-67-5
:
:C₆H₁₃N
:MFCD00006491
:99.18 g/mol
:107°C
:–69 °C
:0,82 g/cm³ (20 °C)
:3°C
:Freezer
:210-959-9
:2399
:3,II
:GHS02
:GHS05
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
Frequently Bought Together