Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
5-tert-Butyl-2-methylthiophenol ≥90.0% (by GC)
  TCIAB1691-25ML
 :  TCI
 :  
undefined
 
 
5-tert-Butyl-2-methylthiophenol ≥90.0% (by GC)
  TCIAB1691-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 90.0 %

 

 

 :
 :180.31 g/mol
 :260 °C (1013 hPa)
 :–10 °C
 :0,98 g/cm³ (20 °C)
 :123°C
 :438-520-9
 :3082
 :9,III
 :GHS07
 :GHS09
 :7340-90-1
 :C₁₁H₁₆S
 :MFCD00142942