
2,4-Dihydroxypyridine ≥98.0% (by GC, titration analysis)
TCIAD2682-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:almost transparency
:
:111.10 g/mol
:315 °C (1013 hPa)
:275…277 °C
:210-924-8
:GHS07
:626-03-9
:84719-31-3
:C₅H₅NO₂
:MFCD00006273