
(S)-(+)-1,2-Propanediol ≥98.0% (by GC)
: TCI
:4254-15-3
:
:C₃H₈O₂
:MFCD00004539
:76.1 g/mol
:14,07855
:92°C
:1,03 g/cm³ (20 °C)
:107°C
:GHS07
:Ambient
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 97.0 ee%
- Specific rotation [a]20/D:+16.0 to +17.0 deg (neat)
Frequently Bought Together