

(3-Mercaptopropyl)trimethoxysilane ≥96.0% (by GC)
: TCI
:4420-74-0
:
:HSH₂CCH₂CH₂Si(OCH₃)₃
:MFCD00004901
:196.34 g/mol
:215 °C (1013 hPa)
:–50 °C
:1,056 g/cm³ (20 °C)
:112°C
:Ambient
:224-588-5
:3082
:9,III
:GHS07
:GHS09
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
- Purity (Iodometric Titration):min. 93.0 %
Frequently Bought Together