Order Entry
Netherlands
ContactUsLinkComponent
6-Methoxy-m-cresol ≥98.0% (by GC)
Irritant
6-Methoxy-m-cresol ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
6-Methoxy-m-cresol ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together