Order Entry
Netherlands
ContactUsLinkComponent
2,7-Dinitrofluorene ≥97.0% (by HPLC, total nitrogen)
2,7-Dinitrofluorene ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
2,7-Dinitrofluorene ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Solubility in hot DMF:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together