Order Entry
Netherlands
ContactUsLinkComponent
4-Nitro-o-toluidine ≥99.0% (by GC)
4-Nitro-o-toluidine ≥99.0% (by GC)
Catalog # TCIAN0529-25G
Supplier:  TCI
CAS Number:  
undefined
4-Nitro-o-toluidine ≥99.0% (by GC)
Catalog # TCIAN0529-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    130.0 to 133.0 deg-C

Specification Test Results

About this item

 :
 :152.15 g/mol
Melting Pt:135 °C
Density:1,366 g/cm³ (15 °C)
Storage Temperature:Ambient
EINECS:202-762-1
UN:2660
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:99-52-5
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007734