
o-Tolyl isothiocyanate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Formula:C₈H₇NS
MDL Number:MFCD00004802
:149.22 g/mol
Flash Pt:107°C
:GHS06
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together