

1,1,1,3,3,3-Hexafluoro-2-propanol ≥99.0% (by GC)
Supplier: Chemicals
Certificates
About this item
CAS Number:920-66-1
:
Formula:C₃H₂F₆O
MDL Number:MFCD00011651
Merck Index:13,09598
Boiling Pt:58°C
Melting Pt:–4°C
Density:1,492 g/cm³ (20 °C)
Flash Pt:≥ 100 °C
UN:1760
ADR:8,III
:168.04 g/mol
EINECS:213-059-4
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together