
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:21963-41-7
:
Formule:C₈H₁₂O₄
MDL-nummer:MFCD00151258
:172.18 g/mol
:GHS07
Temperatuur bij opslag:Kamertemperatuur
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 95.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:180.0 to 185.0 deg-C
Specificatie testresultaten
Frequently Bought Together