
3,3-Dimethylglutaric acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1322-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:102.0 to 105.0 deg-C
- Solubility in Water:very faint turbidity
Specification Test Results
About this item
:
:160.17 g/mol
Boiling Pt:290 °C (1013 hPa)
Melting Pt:98…101 °C
Density:1,4278 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:225-425-0
:GHS07
CAS Number:4839-46-7
Formula:C₇H₁₂O₄
MDL Number:MFCD00002716