Order Entry
Netherlands
ContactUsLinkComponent
5-Aminovaleric acid ≥98.0% (by titrimetric analysis)
5-Aminovaleric acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
5-Aminovaleric acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together