Order Entry
Netherlands
ContactUsLinkComponent
Cesium acetate ≥98.0% (by titrimetric analysis)
Cesium acetate ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
Cesium acetate ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    191.0 to 195.0 deg-C

Specification Test Results

Frequently Bought Together