

1,4-Diamino-9,10-anthraquinone ≥97.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:128-95-0
:
Formula:C₁₄H₁₀N₂O₂
MDL Number:MFCD00001224
:238.25 g/mol
Melting Pt:270°C
EINECS:204-922-6
:GHS07
:GHS08
Specification Test Results
- Appearance:Gray to Dark purple to Black powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
Frequently Bought Together