Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-3-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
Irritant
2-Chloro-3-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
2-Chloro-3-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    169.0 to 173.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together