Order Entry
Netherlands
ContactUsLinkComponent
1,2-Epoxyhexane ≥96.0% (by GC)
Flammable
Irritant
1,2-Epoxyhexane ≥96.0% (by GC)
Catalog # TCIAE0445-25ML
Supplier:  TCI
CAS Number:  
Flammable
Irritant
1,2-Epoxyhexane ≥96.0% (by GC)
Catalog # TCIAE0445-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :100.16 g/mol
Boiling Pt:120°C
Density:0,8456 g/cm³ (20 °C)
Flash Pt:15°C
Storage Temperature:Refrigerator
EINECS:215-864-6
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:1436-34-6
Formula:C₆H₁₂O
MDL Number:MFCD00005154