4-Bromobutyl phenyl ether ≥96.0% (by GC)
Catalog # TCIAP1851-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 96.0 %
- Melting point:39.0 to 43.0 deg-C
Specification Test Results
About this item
:
:229.12 g/mol
Boiling Pt:156°C
Melting Pt:141 °C
EINECS:214-849-1
CAS Number:1200-03-9
Formula:(C₆H₅)O(CH₂)₄Br
MDL Number:MFCD00000262