Bestelling invoeren
Netherlands
ContactUsLinkComponent
 
trans-3-Bromocinnamic acid ≥99.0% (by GC, titration analysis)
 :  TCI
 
undefined
trans-3-Bromocinnamic acid ≥99.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    178.0 to 182.0 deg-C
  • Solubility in Pyridine:
    almost transparency

 

Frequently Bought Together