Order Entry
Netherlands
ContactUsLinkComponent
2,3,5,6-Tetramethylpyrazine ≥98.0% (by GC, titration analysis)
Irritant
2,3,5,6-Tetramethylpyrazine ≥98.0% (by GC, titration analysis)
Catalog # TCIAT0972-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,3,5,6-Tetramethylpyrazine ≥98.0% (by GC, titration analysis)
Catalog # TCIAT0972-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    85.0 to 87.0 deg-C
  • Solubility in Methanol:
    within almost transparency

Specification Test Results

About this item

 :
 :136.2 g/mol
Boiling Pt:190 °C (1013 hPa)
Density:1,15 g/cm³ (20 °C)
EINECS:214-391-2
 :GHS07
Storage Temperature:Ambient
CAS Number:1124-11-4
Formula:C₈H₁₂N₂
MDL Number:MFCD00006146