Order Entry
Netherlands
ContactUsLinkComponent
2,3-Diethylpyrazine ≥98.0% (by GC)
Irritant
2,3-Diethylpyrazine ≥98.0% (by GC)
Catalog # TCIAD1581-5ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,3-Diethylpyrazine ≥98.0% (by GC)
Catalog # TCIAD1581-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :136.2 g/mol
Boiling Pt:71°C
Density:0,97 g/cm³ (20 °C)
Flash Pt:70°C
EINECS:239-800-1
ADR:9,
 :GHS07
CAS Number:15707-24-1
Formula:C₈H₁₂N₂
MDL Number:MFCD00006151