Order Entry
Netherlands
ContactUsLinkComponent
1,3-Diphenylguanidine ≥98.0% (by HPLC, titration analysis)
Irritant
Health
Environment
1,3-Diphenylguanidine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Health
Environment
1,3-Diphenylguanidine ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    147.0 to 152.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together