9,10-Dihydrophenanthrene ≥96.0% (by GC)
: TCI
:776-35-2
:
:C₁₄H₁₂
:MFCD00001164
:180.25 g/mol
:314°C
:34 °C
:1,0953 g/cm³ (14 °C)
:Ambient
:212-278-2
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (GC):min. 96.0 %
- Solubility in Methanol:almost transparency
Frequently Bought Together