1,4-Dihydronaphthalene ≥80.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:612-17-9
:
Formula:C₁₀H₁₀
MDL Number:MFCD00058963
:130.19 g/mol
Boiling Pt:212°C
EINECS:210-297-0
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 80.0 %
Specification Test Results
Frequently Bought Together