



Morpholine ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:110-91-8
:
Formula:C₄H₉NO
MDL Number:MFCD00005972
:87.12 g/mol
Merck Index:14,06277
Boiling Pt:129°C
Melting Pt:–5 °C
Density:1 g/cm³ (20 °C)
Flash Pt:32°C
EINECS:203-815-1
UN:2054
ADR:8,I
:GHS02
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
Specification Test Results
Frequently Bought Together