Order Entry
Netherlands
ContactUsLinkComponent
cis-3-Octene ≥98.0% (by GC)
Flammable
Health
cis-3-Octene ≥98.0% (by GC)
Catalogus # TCIAO0133-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Health
cis-3-Octene ≥98.0% (by GC)
Catalogus # TCIAO0133-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :112.22 g/mol
Boiling Pt:123°C
Melting Pt:–126 °C
Density:0,7159 g/cm³ (20 °C)
Flash Pt:17°C
EINECS:238-912-8
UN:3295
ADR:3,II
 :GHS02
 :GHS08
CAS Number:14850-22-7
Formula:C₈H₁₆
MDL Number:MFCD00067486