

Diphenyl ether ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:101-84-8
:
Formula:C₁₂H₁₀O
MDL Number:MFCD00003034
:170.21 g/mol
Merck Index:14,07288
Boiling Pt:258°C
Melting Pt:24…27 °C
Density:1,07 g/cm³ (20 °C)
Flash Pt:125 °C
Storage Temperature:Ambient
EINECS:202-981-2
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 99.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together