Pyrocatechol ≥99.0% (by GC)
: TCI
:120-80-9
:
:C₆H₆O₂
:MFCD00002188
:110.11 g/mol
:14,07999
:246°C
:103…105 °C
:1,0548 g/cm³ (20 °C)
:127 °C
:Ambient
:204-427-5
:2811
:6.1,III
:GHS05
:GHS06
:GHS08
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 99.0 %
- Solubility in Water:almost transparency
Frequently Bought Together