Pyrocatechol ≥99.0% (by GC), Technical Grade
Supplier: TCI
Certificates
About this item
CAS Number:120-80-9
:
Formula:C₆H₆O₂
MDL Number:MFCD00002188
:110.11 g/mol
Merck Index:14,07999
Boiling Pt:246°C
Melting Pt:103…105 °C
Density:1,0548 g/cm³ (20 °C)
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-427-5
UN:2811
ADR:6.1,III
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:104.0 to 107.0 deg-C
- Solubility in Water:transparency
Specification Test Results
Frequently Bought Together