4(3H)-Pyrimidinone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4562-27-0
:MFCD00167178
Formula:C₄H₄N₂O
:96.09 g/mol
EINECS:224-932-4
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:165.0 to 168.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together