Order Entry
Netherlands
ContactUsLinkComponent
Maleic hydrazide ≥99.0% (by HPLC, titration analysis)
Maleic hydrazide ≥99.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Maleic hydrazide ≥99.0% (by HPLC, titration analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (Neutralization titration):
    min. 99.0 %
  • Solubility in hot Water:
    very faint turbidity

Specification Test Results

Related Information

Frequently Bought Together