

(±)-1-Phenoxy-2-propanol ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:770-35-4
:
Formula:C₆H₅OCH₂CH(OH)CH₃
MDL Number:MFCD00016861
Boiling Pt:233°C
Melting Pt:11.4 °C
Density:1,0665 g/cm³ (20 °C)
Flash Pt:125°C
:152.19 g/mol
Storage Temperature:Ambient
EINECS:212-222-7
:GHS05
:GHS07
Specification Test Results
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together